Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009247.2 | NP_033273.1 | 5 | 1173 | ||
GenBank mRNA | AK004999.1 | - | 5 | 1175 | ||
AK135650.1 | - | 5 | 1179 | |||
AK146576.1 | - | 1375 | ||||
BC061176.1 | - | 5 | 1184 | 30381238 | ||
M75717.1 | - | 5 | 1168 |