Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009210.3 | NP_033236.2 | 7 | 1053 | ||
GenBank mRNA | AB221636.1 | - | 7 | 876 | ||
AF010600.1 | - | 7 | 1053 | |||
AF165911.1 | - | 7 | 1032 | |||
AK141235.1 | - | 7 | 1046 | |||
AK142961.1 | - | 7 | 1010 | |||
AK149897.1 | - | 7 | 1091 | |||
AK166911.1 | - | 7 | 1023 | |||
BC057116.1 | - | 7 | 1022 | 6408672 | ||
BC059240.1 | - | 7 | 1022 | 6408672 |