Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sqle | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009270.3 | NP_033296.1 | 9 | 2168 | ||
GenBank mRNA | AK041198.1 | - | 2 | 709 | ||
AK138457.1 | - | 9 | 2181 | |||
AK148419.1 | - | 9 | 2153 | |||
AK161837.1 | - | 8 | 2058 | |||
AK163502.1 | - | 9 | 2148 | |||
AK189403.1 | - | 3 | 256 | |||
AK208956.1 | - | 3 | 256 | |||
AK209037.1 | - | 3 | 257 | |||
AK209538.1 | - | 3 | 256 | |||
BC042781.1 | - | 9 | 1425 | 4915911 | ||
BC056361.1 | - | 9 | 2168 | 6816800 | ||
D42048.1 | - | 9 | 2034 |