Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159696.1 | NP_001153168.1 | 3, 4 | 774 | ||
NM_009335.2 | NP_033361.2 | 3, 4 | 833 | |||
GenBank mRNA | AK050973.1 | - | 3, 4 | 751 | ||
AK141211.1 | - | 3, 4 | 735 | |||
AK145166.1 | - | 3, 4 | 783 | |||
AK145603.1 | - | 3, 4 | 728 | |||
BC003778.1 | - | 3, 4 | 707 | 3592762 | ||
KJ463836.1 | - | 3, 4 | 629 | |||
X94694.1 | - | 3, 4 | 812 |