Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB011550.1 | - | 3 | 404 | ||
AK076365.1 | - | 1 | 1331 | |||
AK133474.1 | - | 3 | 372 | |||
AK139088.1 | - | 3 | 610 | |||
AK144793.1 | - | 3 | 588 | |||
AK155268.1 | - | 1 | 1349 | |||
BC015077.1 | - | 3 | 287 | |||
BC145595.1 | - | 2 | 378 | |||
BC145596.1 | - | 3 | 293 | |||
U81490.1 | - | 3 | 344 |