siRNA Details
Gene Symbol: Tcf7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142918.1 | NP_001136390.1 | 15 | 3328 | ||
NM_001142919.1 | NP_001136391.1 | 14 | 3277 | |||
NM_001142920.1 | NP_001136392.1 | 12 | 3202 | |||
NM_001142921.1 | NP_001136393.1 | 12 | 3194 | |||
NM_001142922.1 | NP_001136394.1 | 14 | 3303 | |||
NM_001142923.1 | NP_001136395.1 | 12 | 3179 | |||
NM_001142924.1 | NP_001136396.1 | 12 | 2630 | |||
NM_009333.3 | NP_033359.2 | 13 | 3230 | |||
GenBank mRNA | AK018098.1 | - | 1 | 1063 | ||
AK038720.1 | - | 14 | 2722 | |||
AK048536.1 | - | 12 | 2443 | |||
AK052583.1 | - | 8 | 1941 | |||
BC011397.1 | - | 12 | 3157 | |||
BC052022.1 | - | 13 | 2808 | 5702449 |

More Information
Other Species Targeted

Panther Classification:
