Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289550.1 | NP_001276479.1 | 10 | 1243 | ||
NM_001289551.1 | NP_001276480.1 | 10 | 1239 | |||
NM_001289552.1 | NP_001276481.1 | 9 | 1100 | |||
NM_001289553.1 | NP_001276482.1 | 9 | 1127 | |||
GenBank mRNA | AK003170.1 | - | 10 | 1077 | ||
AK041233.1 | - | 10 | 1072 | |||
AK087114.1 | - | 10 | 1074 | |||
AK156423.1 | - | 10 | 1078 | |||
AK158409.1 | - | 9 | 952 | |||
BC002049.1 | - | 10 | 1077 | 3489459 | ||
BC056362.1 | - | 9 | 2352 | 6816930 | ||
DQ143891.1 | - | 9 | 1128 | |||
DQ143892.1 | - | 8 | 914 | |||
DQ143893.1 | - | 8 | 921 | |||
DQ143894.1 | - | 8 | 916 | |||
DQ143895.1 | - | 8 | 1332 | |||
DQ143896.1 | - | 8 | 1293 | |||
DQ143897.1 | - | 8 | 1371 | |||
DQ143898.1 | - | 8 | 1320 | |||
DQ143899.1 | - | 7 | 1447 | |||
DQ143900.1 | - | 9 | 1140 | |||
L22482.1 | - | 9 | 1204 |