Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009373.3 | NP_033399.1 | 13 | 2128 | ||
GenBank mRNA | AF076928.1 | - | 12 | 2043 | ||
AF114266.1 | - | 13 | 2093 | |||
AK052912.1 | - | 13 | 2103 | |||
AK080224.1 | - | 13 | 2128 | |||
AK080593.1 | - | 13 | 2103 | |||
AK086001.1 | - | 13 | 2105 | |||
AK089481.1 | - | 13 | 2131 | |||
AK143712.1 | - | 13 | 2164 | |||
AK151776.1 | - | 13 | 2129 | |||
AK152152.1 | - | 13 | 2103 | |||
AK152627.1 | - | 13 | 2129 | |||
AK159255.1 | - | 13 | 2103 | |||
AK166302.1 | - | 13 | 2103 | |||
AK168990.1 | - | 13 | 2104 | |||
AK169356.1 | - | 13 | 2131 | |||
BC016492.1 | - | 13 | 2069 | 3256943 | ||
M55154.1 | - | 13 | 2062 |