Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT11 SIRNA Assays ›
          Gene Symbol:
          Wnt11
          Gene Name:
          wingless-type MMTV integration site family, member 11
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 98834921 - 98854747 on Build GRCm38
          UniGene ID:
          Mm.22182
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 65214

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Wnt11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001285792.1 NP_001272721.1 5 1036
          NM_001285794.1 NP_001272723.1 4 794
          NM_009519.2 NP_033545.1 3 802
          GenBank mRNA AK031573.1 - 3 646
          AK077828.1 - 4 733
          AK138967.1 - 4 795
          BC125482.1 - 3 749 40129997
          BC125484.1 - 3 749 40129999
          X70800.1 - 3 809

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_004626, BT019492, BC074790, BC113388, AK313570, and others.
          This siRNA also targets the following transcripts in Rat: NM_080401, XM_006229719, XM_006229720, XM_006229718, AB073722.

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          kidney development
          outflow tract morphogenesis
          protein phosphorylation
          signal transduction
          cell-cell signaling
          multicellular organism development
          organ morphogenesis
          positive regulation of gene expression
          Wnt signaling pathway
          neuron differentiation
          bone mineralization
          negative regulation of cell growth
          positive regulation of cell migration
          negative regulation of cell migration
          positive regulation of transforming growth factor beta2 production
          protein localization to cell surface
          non-canonical Wnt signaling pathway
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          cell fate commitment
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          artery morphogenesis
          positive regulation of stress fiber assembly
          palate development
          canonical Wnt signaling pathway
          ventricular septum morphogenesis
          negative regulation of cell death
          neuroendocrine cell differentiation
          bicellular tight junction assembly
          negative regulation of mesenchymal cell proliferation
          positive regulation of protein kinase C signaling
          negative regulation of canonical Wnt signaling pathway
          negative regulation of fibroblast growth factor production
          GTPase activator activity
          receptor binding
          frizzled binding
          protein binding
          protein kinase activator activity
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0