Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNFSF4 SIRNA Assays ›
Gene Symbol:
Tnfsf4
Gene Name:
tumor necrosis factor (ligand) superfamily, member 4
Gene Aliases:
Ath-1, Ath1, CD134L, OX-40L, Ox40l, TXGP1, Txgp1l, gp34
Chromosome Location:
Chr.1: 161395438 - 161418206 on Build GRCm38
UniGene ID:
Mm.4994
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
65252
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnfsf4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009452.2
NP_033478.1
2
320
GenBank mRNA
FJ176943.1
-
2
176
U12763.1
-
2
323
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
cytokine production
defense response to nematode
acute inflammatory response
positive regulation of immune effector process
positive regulation of T cell cytokine production
regulation of adaptive immune response
positive regulation of type 2 immune response
positive regulation of immunoglobulin mediated immune response
inflammatory response
immune response
cholesterol metabolic process
response to virus
negative regulation of interferon-gamma production
negative regulation of interleukin-17 production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-13 production
positive regulation of interleukin-2 production
positive regulation of interleukin-4 production
positive regulation of interleukin-6 production
memory T cell activation
T-helper 2 cell activation
response to nitrogen dioxide
cellular response to nitrogen dioxide
CD4-positive, alpha-beta T cell costimulation
T cell proliferation
positive regulation of activated T cell proliferation
positive regulation of CD4-positive, alpha-beta T cell differentiation
positive regulation of memory T cell differentiation
negative regulation of sequence-specific DNA binding transcription factor activity
innate immune response
negative regulation of regulatory T cell differentiation
negative regulation of T-helper 1 cell differentiation
positive regulation of T-helper 2 cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of alpha-beta T cell proliferation
negative regulation of cytokine secretion
regulation of inflammatory response
positive regulation of inflammatory response
positive regulation of B cell activation
positive regulation of immunoglobulin secretion
negative regulation of T cell apoptotic process
negative regulation of activation-induced cell death of T cells
cellular response to lipopolysaccharide
cellular response to prostaglandin E stimulus
chemokine (C-C motif) ligand 11 production
positive regulation of CD4-positive, alpha-beta T cell costimulation
positive regulation of T cell migration
positive regulation of T cell costimulation
positive regulation of memory T cell activation
positive regulation of T-helper 2 cell activation
positive regulation of interleukin-4-dependent isotype switching to IgE isotypes
negative regulation of extrinsic apoptotic signaling pathway
cytokine activity
tumor necrosis factor receptor binding
tumor necrosis factor receptor superfamily binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0