Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293561.1 | NP_001280490.1 | 2 | 467 | ||
NM_009472.4 | NP_033498.1 | 2 | 467 | |||
GenBank mRNA | AK031655.1 | - | 2 | 434 | ||
AK034558.1 | - | 2 | 434 | |||
AK035038.1 | - | 2 | 433 | |||
AK141181.1 | - | 2 | 433 | |||
AK142411.1 | - | 2 | 433 | |||
AK142974.1 | - | 2 | 361 | |||
BC115772.1 | - | 2 | 457 | 40085998 | ||
U72634.1 | - | 2 | 467 |