Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VCP SIRNA Assays ›
          Gene Symbol:
          Vcp
          Gene Name:
          valosin containing protein
          Gene Aliases:
          3110001E05, CDC48, p97, p97/VCP
          Chromosome Location:
          Chr.4: 42979964 - 43000507 on Build GRCm38
          UniGene ID:
          Mm.245976
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 65291

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Vcp
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009503.4 NP_033529.3 3 518
          GenBank mRNA AK028264.1 - 3 447
          AK030751.1 - 3 349
          AK083821.1 - 1 24
          AK146109.1 - 3 415
          AK149931.1 - 3 411
          AK151109.1 - 3 412
          AK151418.1 - 3 412
          AK153012.1 - 3 448
          AK153249.1 - 3 489
          AK159177.1 - 3 447
          AK159475.1 - 3 446
          AK159509.1 - 3 490
          AK160889.1 - 3 335
          AK167794.1 - 3 466
          AK169140.1 - 3 420
          AK190461.1 - 3 206
          AK194156.1 - 3 206
          AK194828.1 - 3 206
          AK194951.1 - 3 207
          AK194993.1 - 3 206
          AK197087.1 - 3 206
          AK197232.1 - 3 206
          AK199977.1 - 3 206
          AK201680.1 - 3 207
          AK202627.1 - 3 206
          AK202683.1 - 3 206
          AK205885.1 - 3 206
          AK211805.1 - 3 206
          AK212799.1 - 3 206
          AK215162.1 - 3 206
          AK218422.1 - 3 206
          BC043053.1 - 3 422 6418662
          BC049114.1 - 3 466 6810751
          BY732145.1 - 3 518
          Z14044.1 - 3 350

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          double-strand break repair
          ubiquitin-dependent protein catabolic process
          transport
          ER to Golgi vesicle-mediated transport
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to DNA damage stimulus
          protein ubiquitination
          protein N-linked glycosylation via asparagine
          viral genome replication
          translesion synthesis
          ER-associated ubiquitin-dependent protein catabolic process
          retrograde protein transport, ER to cytosol
          positive regulation of protein complex assembly
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein hexamerization
          positive regulation of protein catabolic process
          protein homooligomerization
          aggresome assembly
          ER-associated misfolded protein catabolic process
          positive regulation of protein K63-linked deubiquitination
          positive regulation of Lys63-specific deubiquitinase activity
          nucleotide binding
          receptor binding
          protein binding
          ATP binding
          lipid binding
          hydrolase activity
          ATPase activity
          protein phosphatase binding
          protein domain specific binding
          polyubiquitin binding
          protein complex binding
          deubiquitinase activator activity
          identical protein binding
          ADP binding
          ubiquitin-like protein ligase binding
          poly(A) RNA binding
          ubiquitin-specific protease binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pwwgk:80/100.66.74.89:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0