Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT7A SIRNA Assays ›
          Gene Symbol:
          Wnt7a
          Gene Name:
          wingless-type MMTV integration site family, member 7A
          Gene Aliases:
          AI849442, Wnt-7a, px, tw
          Chromosome Location:
          Chr.6: 91363983 - 91411369 on Build GRCm38
          UniGene ID:
          Mm.56964
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 65314

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Wnt7a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009527.3 NP_033553.2 4 847
          GenBank mRNA AK004683.1 - 4 848
          BC049093.1 - 4 841 6415801
          BC057586.1 - 4 841 6415801
          M89801.1 - 4 587

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          cartilage condensation
          angiogenesis
          chondrocyte differentiation
          signal transduction
          cell-cell signaling
          neurotransmitter secretion
          multicellular organism development
          asymmetric protein localization
          positive regulation of cell proliferation
          organ morphogenesis
          dorsal/ventral pattern formation
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          skeletal muscle satellite cell activation
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          Wnt signaling pathway
          cerebellar granule cell differentiation
          cell proliferation in forebrain
          central nervous system vasculogenesis
          establishment of cell polarity
          embryonic limb morphogenesis
          regulation of axon diameter
          somatic stem cell population maintenance
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          forelimb morphogenesis
          hindlimb morphogenesis
          non-canonical Wnt signaling pathway
          Wnt signaling pathway involved in wound healing, spreading of epidermal cells
          regulation of cell proliferation
          embryonic digit morphogenesis
          negative regulation of apoptotic process
          skin morphogenesis
          response to estrogen
          cell fate commitment
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          somatic stem cell division
          reproductive structure development
          skeletal system morphogenesis
          stem cell development
          negative regulation of neurogenesis
          regulation of axonogenesis
          synapse organization
          cartilage development
          positive regulation of synapse assembly
          palate development
          positive regulation of epithelial cell proliferation involved in wound healing
          uterus development
          oviduct development
          canonical Wnt signaling pathway
          limb development
          uterus morphogenesis
          positive regulation of canonical Wnt signaling pathway
          positive regulation of receptor activity
          receptor binding
          frizzled binding
          cytokine activity
          protein binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cdcg2:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0