Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNFSF4 SIRNA Assays ›
          Gene Symbol:
          Tnfsf4
          Gene Name:
          tumor necrosis factor (ligand) superfamily, member 4
          Gene Aliases:
          Ath-1, Ath1, CD134L, OX-40L, Ox40l, TXGP1, Txgp1l, gp34
          Chromosome Location:
          Chr.1: 161395438 - 161418206 on Build GRCm38
          UniGene ID:
          Mm.4994
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 65342

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tnfsf4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009452.2 NP_033478.1 3 1134
          GenBank mRNA U12763.1 - 3 1135

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          cytokine production
          defense response to nematode
          acute inflammatory response
          positive regulation of immune effector process
          positive regulation of T cell cytokine production
          regulation of adaptive immune response
          positive regulation of type 2 immune response
          positive regulation of immunoglobulin mediated immune response
          inflammatory response
          immune response
          cholesterol metabolic process
          response to virus
          negative regulation of interferon-gamma production
          negative regulation of interleukin-17 production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-13 production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-4 production
          positive regulation of interleukin-6 production
          memory T cell activation
          T-helper 2 cell activation
          response to nitrogen dioxide
          cellular response to nitrogen dioxide
          CD4-positive, alpha-beta T cell costimulation
          T cell proliferation
          positive regulation of activated T cell proliferation
          positive regulation of CD4-positive, alpha-beta T cell differentiation
          positive regulation of memory T cell differentiation
          negative regulation of sequence-specific DNA binding transcription factor activity
          innate immune response
          negative regulation of regulatory T cell differentiation
          negative regulation of T-helper 1 cell differentiation
          positive regulation of T-helper 2 cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of alpha-beta T cell proliferation
          negative regulation of cytokine secretion
          regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of B cell activation
          positive regulation of immunoglobulin secretion
          negative regulation of T cell apoptotic process
          negative regulation of activation-induced cell death of T cells
          cellular response to lipopolysaccharide
          cellular response to prostaglandin E stimulus
          chemokine (C-C motif) ligand 11 production
          positive regulation of CD4-positive, alpha-beta T cell costimulation
          positive regulation of T cell migration
          positive regulation of T cell costimulation
          positive regulation of memory T cell activation
          positive regulation of T-helper 2 cell activation
          positive regulation of interleukin-4-dependent isotype switching to IgE isotypes
          negative regulation of extrinsic apoptotic signaling pathway
          cytokine activity
          tumor necrosis factor receptor binding
          tumor necrosis factor receptor superfamily binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0