Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AXL SIRNA Assays ›
          Gene Symbol:
          Axl
          Gene Name:
          AXL receptor tyrosine kinase
          Gene Aliases:
          AI323647, Ark, Tyro7, Ufo
          Chromosome Location:
          Chr.7: 25756500 - 25788733 on Build GRCm38
          UniGene ID:
          Mm.4128
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 65354

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Axl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190974.1 NP_001177903.1 19 3118
          NM_001190975.1 NP_001177904.1 18 2854
          NM_009465.4 NP_033491.2 20 3145
          GenBank mRNA AK048382.1 - 1 511
          AK155567.1 - 21 3031
          AK161294.1 - 13 1818
          AK170010.1 - 19 3092
          AK170897.1 - 19 2753
          BC008551.1 - 15 2235
          BC046618.1 - 20 2982 6494383
          BC050914.1 - 20 2953 6418252
          BC058230.1 - 20 3006 6313662
          X63535.1 - 20 2894

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          natural killer cell differentiation
          positive regulation of cytokine-mediated signaling pathway
          blood vessel remodeling
          immune system process
          protein phosphorylation
          phagocytosis
          inflammatory response
          enzyme linked receptor protein signaling pathway
          spermatogenesis
          phosphorylation
          peptidyl-tyrosine phosphorylation
          forebrain cell migration
          cell differentiation
          platelet activation
          cellular response to extracellular stimulus
          negative regulation of interferon-gamma production
          negative regulation of tumor necrosis factor production
          positive regulation of natural killer cell differentiation
          secretion by cell
          erythrocyte homeostasis
          substrate adhesion-dependent cell spreading
          cellular response to interferon-alpha
          ovulation cycle
          negative regulation of apoptotic process
          apoptotic cell clearance
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          innate immune response
          viral entry into host cell
          positive regulation of pinocytosis
          negative regulation of lymphocyte activation
          positive regulation of protein kinase B signaling
          vagina development
          cellular response to hydrogen peroxide
          cellular response to lipopolysaccharide
          negative regulation of dendritic cell apoptotic process
          nucleotide binding
          phosphatidylserine binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          myosin heavy chain binding
          phosphatidylinositol 3-kinase binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0