Search
Search

Zfp57| Gene Symbol: Zfp57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013745.2 | NP_001013767.1 | 5 | 793 | ||
| NM_001168501.1 | NP_001161973.1 | 5 | 659 | |||
| NM_001168502.1 | NP_001161974.1 | 6 | 1035 | |||
| GenBank mRNA | AK054083.1 | - | 5 | 776 | ||
| AK135881.1 | - | 5 | 767 | |||
| AK136241.1 | - | 5 | 777 | |||
| AK139625.1 | - | 6 | 1035 | |||
| AK143118.1 | - | 3 | 1641 | |||
| AK145368.1 | - | 5 | 765 | |||
| BC052028.1 | - | 3 | 960 | 5686931 | ||
| D21850.1 | - | 4 | 788 | |||