siRNA Details
Gene Symbol: Aqp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009699.3 | NP_033829.3 | 3 | 679 | ||
GenBank mRNA | AF020519.1 | - | 3 | 582 | ||
AK002689.1 | - | 3 | 680 | |||
AK142567.1 | - | 3 | 680 | |||
BC019966.1 | - | 3 | 663 | 4222942 |
