Search
Search

Aspm| Gene Symbol: Aspm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009791.4 | NP_033921.3 | 19 | 7990 | ||
| GenBank mRNA | AF062378.1 | - | 2 | 250 | ||
| AF533752.1 | - | 19 | 7968 | |||
| AK048277.1 | - | 2 | 802 | |||
| AY971958.1 | - | 18 | 4036 | |||