Search
Search

Aspm| Gene Symbol: Aspm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009791.4 | NP_033921.3 | 21 | 8315 | ||
| GenBank mRNA | AF062378.1 | - | 4 | 575 | ||
| AF533752.1 | - | 21 | 8293 | |||
| AK048277.1 | - | 4 | 1127 | |||
| AY971958.1 | - | 20 | 4361 | |||