siRNA Details
Gene Symbol: GAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005255.3 | NP_005246.2 | 3 | 439 | ||
XM_005272268.2 | XP_005272325.1 | 3 | 379 | |||
XM_005272270.2 | XP_005272327.1 | 3 | 379 | |||
XM_011513425.2 | XP_011511727.1 | 3 | 379 | |||
XM_011513426.2 | XP_011511728.1 | 3 | 379 | |||
XM_011513427.2 | XP_011511729.1 | 3 | 379 | |||
XM_011513428.2 | XP_011511730.1 | 3 | 379 | |||
XM_011513431.2 | XP_011511733.1 | 3 | 379 | |||
XM_011513432.2 | XP_011511734.1 | 3 | 350 | |||
XM_017007991.1 | XP_016863480.1 | 3 | 379 | |||
XM_017007994.1 | XP_016863483.1 | 3 | 379 | |||
XM_017007995.1 | XP_016863484.1 | 3 | 341 | |||
GenBank mRNA | BC001290.1 | - | 3 | 414 | ||
BC063606.1 | - | 3 | 343 | |||
D88435.1 | - | 3 | 236 | |||
DA680073.1 | - | 3 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -





