Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELSR1 SIRNA Assays ›
Gene Symbol:
Celsr1
Gene Name:
cadherin, EGF LAG seven-pass G-type receptor 1 (flamingo homolog, Drosophila)
Gene Aliases:
Adgrc1, Crsh, Scy, crash
Chromosome Location:
Chr.15: 85898758 - 86034217 on Build GRCm38
UniGene ID:
Mm.22680
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
66135
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Celsr1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009886.2
NP_034016.2
1
1008
GenBank mRNA
AF031572.1
-
1
1008
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006242167, XM_008765821, XM_008765820.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
gastrulation with mouth forming second
establishment of planar polarity
neuron migration
neural tube closure
hair follicle development
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
multicellular organism development
central nervous system development
locomotory behavior
anterior/posterior pattern specification
regulation of actin cytoskeleton organization
wound healing
establishment of planar polarity of embryonic epithelium
inner ear morphogenesis
apical protein localization
regulation of neurotransmitter secretion
establishment of body hair planar orientation
orthogonal dichotomous subdivision of terminal units involved in lung branching morphogenesis
planar dichotomous subdivision of terminal units involved in lung branching morphogenesis
lateral sprouting involved in lung morphogenesis
planar cell polarity pathway involved in neural tube closure
protein localization involved in establishment of planar polarity
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding
protein binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0