Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAT SIRNA Assays ›
          Gene Symbol:
          Cat
          Gene Name:
          catalase
          Gene Aliases:
          2210418N07, Cas-1, Cas1, Cs-1
          Chromosome Location:
          Chr.2: 103453904 - 103485153 on Build GRCm38
          UniGene ID:
          Mm.4215
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 66163

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cat
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009804.2 NP_033934.2 3 417
          GenBank mRNA AK075853.1 - 3 387
          AK088168.1 - 3 379
          AK128919.1 - 3 392
          AK133881.1 - 3 385
          AK148961.1 - 3 385
          AK150893.1 - 3 388
          AK159152.1 - 3 388
          AK159885.1 - 3 388
          AK159891.1 - 3 339
          AK159913.1 - 3 369
          AK169069.1 - 3 382
          AK188260.1 - 2 101
          AK188401.1 - 2 101
          AY040626.1 - 3 358
          BC013447.1 - 3 374 4241333
          BC047126.1 - 3 332
          L25069.1 - 3 389
          M62897.1 - 3 302
          X52108.1 - 3 302

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          osteoblast differentiation
          ureteric bud development
          response to hypoxia
          kidney development
          triglyceride metabolic process
          response to oxidative stress
          cholesterol metabolic process
          aerobic respiration
          UV protection
          positive regulation of phosphatidylinositol 3-kinase signaling
          hemoglobin metabolic process
          negative regulation of NF-kappaB transcription factor activity
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          positive regulation of NF-kappaB transcription factor activity
          protein tetramerization
          protein homotetramerization
          positive regulation of cell division
          oxidation-reduction process
          aminoacylase activity
          catalase activity
          peroxidase activity
          receptor binding
          antioxidant activity
          oxidoreductase activity
          oxidoreductase activity, acting on peroxide as acceptor
          enzyme binding
          heme binding
          protein homodimerization activity
          metal ion binding
          NADP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lj6zv:80/100.66.79.197:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0