Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001755.2 | NP_001746.1 | 6 | 1227 | ||
NM_022845.2 | NP_074036.1 | 6 | 1196 | |||
XM_005256212.3 | XP_005256269.1 | 5 | 1041 | |||
XM_006721321.2 | XP_006721384.1 | 5 | 1041 | |||
XM_017023812.1 | XP_016879301.1 | 5 | 1072 | |||
XM_017023813.1 | XP_016879302.1 | 5 | 1096 | |||
GenBank mRNA | AK290462.1 | - | 6 | 985 | ||
AK311084.1 | - | 2 | 554 | |||
AK311724.1 | - | 3 | 552 | |||
BC018509.1 | - | 6 | 1227 | 3867061 | ||
L20298.1 | - | 6 | 945 |