Search
Search

Csnk2a2| Gene Symbol: Csnk2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009974.3 | NP_034104.1 | 12 | 1700 | ||
| GenBank mRNA | AF012251.1 | - | 12 | 1425 | ||
| AJ001420.1 | - | 12 | 1669 | |||
| AK002927.1 | - | 12 | 1708 | |||
| AK030035.1 | - | 6 | 740 | |||
| BC047143.1 | - | 12 | 1690 | |||
| BC057862.1 | - | 12 | 1360 | 5316132 | ||