Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110228.1 | NP_001103698.1 | 2 | 262 | ||
NM_001110229.1 | NP_001103699.1 | 3 | 349 | |||
NM_001110230.1 | NP_001103700.1 | 3 | 349 | |||
NM_001110231.1 | NP_001103701.1 | 5 | 324 | |||
NM_001110232.1 | NP_001103702.1 | 4 | 335 | |||
NM_001160292.1 | NP_001153764.1 | 3 | 349 | |||
NM_001160293.1 | NP_001153765.1 | 2 | 262 | |||
NM_010160.2 | NP_034290.2 | 2 | 262 | |||
GenBank mRNA | AF090696.1 | - | 4 | 335 | ||
AF090697.1 | - | 2 | 235 | |||
AK037436.1 | - | 1 | 721 | |||
AK135166.1 | - | 2 | 213 | |||
AK137292.1 | - | 3 | 290 | |||
AK137555.1 | - | 1 | 120 | |||
AK151818.1 | - | 1 | 120 | |||
AK152628.1 | - | 1 | 120 | |||
AK153267.1 | - | 1 | 120 | |||
AK159996.1 | - | 3 | 356 | |||
AK160861.1 | - | 5 | 325 | |||
AK162651.1 | - | 1 | 120 | |||
AK164112.1 | - | 2 | 159 | |||
AK164914.1 | - | 3 | 290 | |||
AK169542.1 | - | 1 | 135 | |||
AK172648.1 | - | 1 | 120 | |||
AK193426.1 | - | 2 | 291 | |||
AK213125.1 | - | 2 | 185 | |||
AK219050.1 | - | 1 | 67 | |||
BC026856.1 | - | 2 | 213 | 4503295 | ||
BY145030.1 | - | 2 | 219 | |||
CF536042.1 | - | 2 | 262 | |||
Y18298.1 | - | 1 | 128 |