Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GNAS SIRNA Assays ›
Gene Symbol:
GNAS
Gene Name:
GNAS complex locus
Gene Aliases:
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
Chromosome Location:
Chr.20: 58839681 - 58911196 on Build GRCh38
UniGene ID:
Hs.125898
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6673
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GNAS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000516.5
NP_000507.1
2
614
NM_001077488.3
NP_001070956.1
2
614
NM_001077489.3
NP_001070957.1
2
614
NM_001077490.2
NP_001070958.1
2
2404
NM_001309840.1
NP_001296769.1
2
280
NM_001309842.1
NP_001296771.1
2
614
NM_001309861.1
NP_001296790.1
2
325
NM_001309883.1
NP_001296812.1
4
2562
NM_016592.3
NP_057676.1
2
1220
NM_080425.3
NP_536350.2
2
2404
NM_080426.3
NP_536351.1
2
614
NR_132272.1
2
97
NR_132273.1
2
457
XM_017027812.1
XP_016883301.1
2
2671
XM_017027813.1
XP_016883302.1
2
2671
XM_017027814.1
XP_016883303.1
2
2671
XM_017027815.1
XP_016883304.1
2
142
XM_017027816.1
XP_016883305.1
2
820
XM_017027817.1
XP_016883306.1
2
229
XM_017027818.1
XP_016883307.1
2
152
XM_017027819.1
XP_016883308.1
2
193
XM_017027820.1
XP_016883309.1
1
51
XM_017027821.1
XP_016883310.1
2
1257
XM_017027822.1
XP_016883311.1
2
1257
GenBank mRNA
AF105253.1
-
2
1198
AF493897.1
-
2
190
AF493898.1
-
2
190
AJ224867.1
-
4
585
AK026564.1
-
3
384
AK093534.1
-
1
466
AK225818.1
-
3
358
AK315860.1
-
2
112
AK315874.1
-
2
293
AL537118.3
-
2
310
AV751821.1
-
2
457
BC002722.2
-
3
308
BC008855.2
-
3
332
BC022875.1
-
2
81
BC066923.1
-
2
193
BC089157.1
-
2
506
BC104928.1
-
3
278
BC108315.1
-
2
455
BM894355.1
-
2
155
BT009905.1
-
2
190
BU857975.1
-
2
504
CN295544.1
-
2
278
CN403086.1
-
2
238
DB483472.1
-
2
193
HY141824.1
-
2
97
KU178095.1
-
2
190
KU178096.1
-
2
190
M14631.1
-
2
157
X04408.1
-
2
258
X04409.1
-
2
202
X07036.1
-
2
202
X56009.1
-
2
266
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_201618, NM_001077507, NM_019690, NM_010309, NM_001077510, and others.
Panther Classification:
Molecular Function -
heterotrimeric G-protein
Gene Ontology Categories:
Function(s)
Process(es)
tissue homeostasis
endochondral ossification
energy reserve metabolic process
DNA methylation
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-activating dopamine receptor signaling pathway
sensory perception of chemical stimulus
embryonic hindlimb morphogenesis
multicellular organism growth
post-embryonic body morphogenesis
response to drug
positive regulation of osteoblast differentiation
positive regulation of osteoclast differentiation
developmental growth
embryonic cranial skeleton morphogenesis
cognition
cartilage development
bone development
hair follicle placode formation
platelet aggregation
genetic imprinting
regulation of parathyroid hormone secretion
GTPase activity
signal transducer activity
insulin-like growth factor receptor binding
protein binding
GTP binding
G-protein beta/gamma-subunit complex binding
beta-2 adrenergic receptor binding
D1 dopamine receptor binding
mu-type opioid receptor binding
ionotropic glutamate receptor binding
metal ion binding
corticotropin-releasing hormone receptor 1 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0