Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010112.4 | NP_034242.2 | 2, 3 | 1635 | ||
GenBank mRNA | AK030321.1 | - | 2, 3 | 762 | ||
AK163191.1 | - | 2, 3 | 676 | |||
BC005438.1 | - | 2, 3 | 704 | 3487600 | ||
BC031543.1 | - | 2, 3 | 430 | 3669513 | ||
U28728.1 | - | 2, 3 | 659 | |||
U57686.1 | - | 2, 3 | 359 |