Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271470.1 | NP_001258399.1 | 9 | 1411 | ||
NM_010249.2 | NP_034379.1 | 8 | 1249 | |||
GenBank mRNA | AK139710.1 | - | 9 | 1411 | ||
AY282803.1 | - | 5 | 706 | |||
AY282804.1 | - | 6 | 788 | |||
AY282805.1 | - | 5 | 789 | |||
M74517.1 | - | 8 | 1143 |