Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163165.1 | NP_001156637.1 | 5 | 546 | ||
NM_001163166.1 | NP_001156638.1 | 5 | 443 | |||
NM_010440.3 | NP_034570.1 | 4 | 630 | |||
GenBank mRNA | AF146224.1 | - | 4 | 613 | ||
AK153816.1 | - | 5 | 444 | |||
AK155615.1 | - | 5 | 547 | |||
AK155652.1 | - | 5 | 448 | |||
AK155890.1 | - | 4 | 506 | |||
AK183154.1 | - | 1 | 23 | |||
AK186994.1 | - | 1 | 23 | |||
AK189436.1 | - | 1 | 23 | |||
AK216551.1 | - | 1 | 23 | |||
AL355734.1 | - | 4 | 625 | |||
BC003924.1 | - | 4 | 518 | 3485302 | ||
BC011334.1 | - | 5 | 479 | 4012515 | ||
BC023436.1 | - | 2 | 106 | |||
BC071217.1 | - | 2 | 1507 |