Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010474.2 | NP_034604.1 | 2 | 436 | ||
GenBank mRNA | AF019385.1 | - | 437 | |||
AK087753.1 | - | 2 | 415 | |||
BC009133.1 | - | 2 | 398 | 3155049 | ||
CJ144446.1 | - | 2 | 439 |