Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLR4 SIRNA Assays ›
          Gene Symbol:
          TLR4
          Gene Name:
          toll like receptor 4
          Gene Aliases:
          ARMD10, CD284, TLR-4, TOLL
          Chromosome Location:
          Chr.9: 117704175 - 117717491 on Build GRCh38
          UniGene ID:
          Hs.174312
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 6786

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLR4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003266.3 NP_003257.1 4 1225
          NM_138554.4 NP_612564.1 3 1105
          NM_138557.2 NP_612567.1 2 938
          GenBank mRNA AB445638.1 - 3 807
          AK226155.1 - 4 980
          AK290053.1 - 4 1208
          AK293068.1 - 3 936
          AK303730.1 - 2 769
          BC025294.1 - 2 724
          BC117422.1 - 3 827 40125973
          BC143841.1 - 4 947
          U88880.1 - 4 971
          U93091.1 - 3 910

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          toll-like receptor signaling pathway
          B cell proliferation involved in immune response
          nitric oxide production involved in inflammatory response
          regulation of dendritic cell cytokine production
          MyD88-dependent toll-like receptor signaling pathway
          MyD88-independent toll-like receptor signaling pathway
          inflammatory response
          immune response
          I-kappaB kinase/NF-kappaB signaling
          I-kappaB phosphorylation
          positive regulation of platelet activation
          positive regulation of gene expression
          astrocyte development
          detection of fungus
          positive regulation of B cell proliferation
          lipopolysaccharide-mediated signaling pathway
          response to lipopolysaccharide
          detection of lipopolysaccharide
          interferon-gamma production
          negative regulation of interferon-gamma production
          negative regulation of interleukin-17 production
          negative regulation of interleukin-23 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of chemokine production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          negative regulation of MyD88-independent toll-like receptor signaling pathway
          toll-like receptor 4 signaling pathway
          TRIF-dependent toll-like receptor signaling pathway
          T-helper 1 type immune response
          macrophage activation
          positive regulation of NF-kappaB import into nucleus
          positive regulation of tumor necrosis factor biosynthetic process
          defense response to bacterium
          positive regulation of interleukin-12 biosynthetic process
          innate immune response
          positive regulation of MHC class II biosynthetic process
          positive regulation of interferon-beta biosynthetic process
          positive regulation of interleukin-8 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          negative regulation of osteoclast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          interleukin-1 beta secretion
          regulation of cytokine secretion
          positive regulation of inflammatory response
          defense response to Gram-negative bacterium
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          intestinal epithelial structure maintenance
          positive regulation of macrophage cytokine production
          necroptotic process
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway
          positive regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to mechanical stimulus
          apoptotic signaling pathway
          positive regulation of NLRP3 inflammasome complex assembly
          lipopolysaccharide binding
          lipopolysaccharide receptor activity
          receptor activity
          transmembrane signaling receptor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0