Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010752.3 | NP_034882.2 | 4 | 462 | ||
GenBank mRNA | AF083812.1 | - | 4 | 469 | ||
AF083813.1 | - | 4 | 469 | |||
AK136981.1 | - | 4 | 464 | |||
BC010702.1 | - | 4 | 460 | 3156639 | ||
BI735882.1 | - | 4 | 461 |