Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CITED2 SIRNA Assays ›
          Gene Symbol:
          Cited2
          Gene Name:
          Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2
          Gene Aliases:
          AI835299, ER154-like, Mrg1, Msg2, p35srj
          Chromosome Location:
          Chr.10: 17723205 - 17725674 on Build GRCm38
          UniGene ID:
          Mm.272321
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68187

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cited2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010828.3 NP_034958.2 2 523
          GenBank mRNA AK131664.1 - 2 526
          AK133791.1 - 2 524
          BC057126.1 - 2 505 6415181
          U86445.1 - 2 503
          Y15163.1 - 2 502

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001168389, NM_006079, NM_001168388, AF109161, U65093, and others.
          This siRNA also targets the following transcripts in Rat: NM_053698, AF361476, BC087005.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          vasculogenesis
          branching involved in ureteric bud morphogenesis
          response to hypoxia
          in utero embryonic development
          kidney development
          trophectodermal cell differentiation
          neural tube formation
          neural tube closure
          liver development
          embryonic placenta development
          vasculature development
          heart looping
          morphogenesis of an epithelium
          lens morphogenesis in camera-type eye
          hematopoietic progenitor cell differentiation
          leukocyte differentiation
          outflow tract morphogenesis
          regulation of organ formation
          endocardial cushion development
          ventricular septum development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          determination of left/right symmetry
          central nervous system development
          peripheral nervous system development
          heart development
          sex determination
          cell proliferation
          male gonad development
          positive regulation of gene expression
          negative regulation of gene expression
          cranial nerve morphogenesis
          positive regulation of cell-cell adhesion
          cell differentiation
          adrenal gland development
          negative regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          granulocyte differentiation
          response to fluid shear stress
          positive regulation of peroxisome proliferator activated receptor signaling pathway
          adrenal cortex formation
          skeletal muscle cell differentiation
          negative regulation of apoptotic process
          response to estrogen
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          decidualization
          spleen development
          thymus development
          embryonic camera-type eye morphogenesis
          erythrocyte development
          embryonic process involved in female pregnancy
          bone morphogenesis
          cardiac septum morphogenesis
          ventricular septum morphogenesis
          embryonic heart tube left/right pattern formation
          pulmonary artery morphogenesis
          cardiac neural crest cell development involved in heart development
          determination of heart left/right asymmetry
          negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
          left/right axis specification
          nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          regulation of nucleic acid-templated transcription
          positive regulation of male gonad development
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          RNA polymerase II transcription corepressor activity
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription cofactor activity
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          histone acetyltransferase binding
          SMAD binding
          LBD domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0