Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010218.1 | - | 3 | 380 | ||
AK038631.1 | - | 3 | 428 | |||
AK156731.1 | - | 3 | 354 | |||
AK202557.1 | - | 2 | 116 | |||
AY176058.1 | - | 3 | 387 | |||
AY176059.1 | - | 3 | 418 | |||
BC021938.1 | - | 3 | 385 | 5136809 | ||
U19799.1 | - | 3 | 399 |