Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010892.3 | NP_035022.2 | 7 | 1171 | ||
GenBank mRNA | AF007247.1 | - | 7 | 1085 | ||
AF013166.1 | - | 7 | 1055 | |||
AK147072.1 | - | 7 | 1147 | |||
AK164467.1 | - | 7 | 1114 | |||
AK169330.1 | - | 8 | 1062 | |||
AK170179.1 | - | 7 | 1091 | |||
AK171277.1 | - | 7 | 1091 | |||
AK171387.1 | - | 7 | 1091 | |||
BC010302.1 | - | 7 | 1093 | 3597227 | ||
BC011316.1 | - | 7 | 1091 | 3674001 | ||
BC057576.1 | - | 7 | 1111 | 6810914 | ||
U95610.1 | - | 7 | 1082 |