Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181325.4 | NP_851842.1 | 3 | 352 | ||
GenBank mRNA | AF133914.1 | - | 3 | 200 | ||
AK052758.1 | - | 3 | 284 | |||
AK081243.1 | - | 3 | 275 | |||
AK142223.1 | - | 3 | 222 | |||
AK145966.1 | - | 3 | 222 | |||
AK154988.1 | - | 3 | 328 | |||
BC061143.1 | - | 3 | 202 | 30276598 | ||
CJ172337.1 | - | 3 | 352 |