Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other P2RX7 SIRNA Assays ›
          Gene Symbol:
          P2rx7
          Gene Name:
          purinergic receptor P2X, ligand-gated ion channel, 7
          Gene Aliases:
          AI467586, P2X(7), P2X7R
          Chromosome Location:
          Chr.5: 122643911 - 122691432 on Build GRCm38
          UniGene ID:
          Mm.42026
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68482

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: P2rx7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001038839.2 NP_001033928.1 2 363
          NM_001038845.2 NP_001033934.1 2 363
          NM_001038887.1 NP_001033976.1 2 363
          NM_001284402.1 NP_001271331.1 2 191
          NM_011027.3 NP_035157.2 2 363
          GenBank mRNA AJ009823.1 - 2 211
          AJ489296.1 - 2 200
          AJ489297.1 - 2 200
          AK144585.1 - 2 364
          AK145050.1 - 2 218
          AK145211.1 - 2 227
          AK171132.1 - 2 335
          BC062808.1 - 2 265
          BC128303.1 - 2 207
          BC128304.1 - 2 207
          BC141120.1 - 2 230 9055732
          BY752747.1 - 2 305
          FJ436444.1 - 2 191

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          cell morphogenesis
          phagolysosome assembly
          positive regulation of T cell mediated cytotoxicity
          positive regulation of protein phosphorylation
          protein phosphorylation
          membrane protein ectodomain proteolysis
          phospholipid transfer to membrane
          transport
          ion transport
          cation transport
          calcium ion transport
          cell volume homeostasis
          membrane budding
          inflammatory response
          mitochondrion organization
          plasma membrane organization
          response to mechanical stimulus
          response to bacterium
          response to organic substance
          response to zinc ion
          gene expression
          positive regulation of calcium ion transport into cytosol
          positive regulation of gene expression
          programmed cell death
          positive regulation of glutamate secretion
          positive regulation of gamma-aminobutyric acid secretion
          response to organic cyclic compound
          synaptic vesicle exocytosis
          protein processing
          phospholipid scrambling
          sensory perception of pain
          cytolysis
          positive regulation of bone mineralization
          cellular response to extracellular stimulus
          bleb assembly
          positive regulation of prostaglandin secretion
          response to lipopolysaccharide
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-6 production
          collagen metabolic process
          response to ATP
          response to fluid shear stress
          positive regulation of ion transmembrane transport
          purinergic nucleotide receptor signaling pathway
          T cell proliferation
          response to drug
          T cell homeostasis
          positive regulation of apoptotic process
          positive regulation of catalytic activity
          NAD transport
          negative regulation of MAPK cascade
          positive regulation of MAPK cascade
          multicellular organismal protein catabolic process
          phospholipid translocation
          positive regulation of ossification
          negative regulation of bone resorption
          negative regulation of cell volume
          positive regulation of glycolytic process
          ceramide biosynthetic process
          pore complex assembly
          skeletal system morphogenesis
          homeostasis of number of cells within a tissue
          positive regulation of protein secretion
          positive regulation of cytokine secretion
          positive regulation of interleukin-1 alpha secretion
          positive regulation of interleukin-1 beta secretion
          defense response to Gram-positive bacterium
          release of sequestered calcium ion into cytosol
          protein oligomerization
          response to calcium ion
          response to electrical stimulus
          membrane depolarization
          positive regulation of mitochondrial depolarization
          positive regulation of lymphocyte apoptotic process
          cellular response to dsRNA
          cellular response to organic cyclic compound
          reactive oxygen species metabolic process
          extrinsic apoptotic signaling pathway
          cation transmembrane transport
          positive regulation of bleb assembly
          lipopolysaccharide binding
          purinergic nucleotide receptor activity
          receptor activity
          extracellular ATP-gated cation channel activity
          ion channel activity
          protein binding
          ATP binding
          channel activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0