Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164198.1 | NP_001157670.1 | 3 | 393 | ||
NM_001164199.1 | NP_001157671.1 | 3 | 266 | |||
NM_001164200.1 | NP_001157672.1 | 3 | 358 | |||
NM_011100.4 | NP_035230.1 | 3 | 397 | |||
GenBank mRNA | AK048319.1 | - | 3 | 389 | ||
AK139419.1 | - | 3 | 138 | |||
AK148728.1 | - | 3 | 207 | |||
AK150122.1 | - | 3 | 359 | |||
AK150134.1 | - | 3 | 359 | |||
AK163585.1 | - | 3 | 171 | |||
AK193506.1 | - | 3 | 140 | |||
AK204631.1 | - | 3 | 140 | |||
AK216103.1 | - | 3 | 192 | |||
BC054533.1 | - | 3 | 381 | 5704796 | ||
BY129968.1 | - | 3 | 267 | |||
BY152174.1 | - | 3 | 411 | |||
CB723114.1 | - | 3 | 393 | |||
J02626.1 | - | 3 | 326 | |||
JN189786.1 | - | 3 | 209 | |||
JN189787.1 | - | 3 | 158 | |||
JN189788.1 | - | 3 | 133 |