Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162387.1 | NP_001155859.1 | 4 | 740 | ||
NM_001162388.1 | NP_001155860.1 | 4 | 504 | |||
NM_011069.3 | NP_035199.3 | 4 | 740 | |||
GenBank mRNA | AF093671.1 | - | 4 | 485 | ||
AK005236.1 | - | 4 | 492 | |||
AK011709.1 | - | 4 | 538 | |||
AK082279.1 | - | 4 | 499 | |||
AK082778.1 | - | 4 | 461 | |||
BC013812.1 | - | 4 | 489 | 3483892 | ||
BC054414.1 | - | 4 | 488 | 3257217 | ||
BQ443231.1 | - | 4 | 550 |