Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011228.2 | NP_035358.1 | 1 | 545 | ||
GenBank mRNA | AB232628.1 | - | 1 | 229 | ||
BC104390.1 | - | 1 | 293 | 40050293 | ||
BC104391.1 | - | 1 | 293 | 40050296 | ||
D83279.1 | - | 1 | 360 |