Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282000.1 | NP_001268929.1 | 1, 2 | 358 | ||
NM_001282001.1 | NP_001268930.1 | 1, 2 | 358 | |||
NM_011250.4 | NP_035380.3 | 1, 2 | 358 | |||
GenBank mRNA | AK142172.1 | - | 1, 2 | 317 | ||
AK155256.1 | - | 1, 2 | 322 | |||
AK160027.1 | - | 1, 2 | 305 | |||
BC020528.1 | - | 1, 2 | 306 | 4240592 | ||
BY270001.1 | - | 1, 2 | 358 | |||
BY749910.1 | - | 1, 2 | 322 | |||
U36799.1 | - | 1, 2 | 258 | |||
U47333.1 | - | 1, 2 | 300 | |||
U50850.1 | - | 1, 2 | 266 |