Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284392.1 | NP_001271321.1 | 11 | 1208 | ||
NM_001284394.1 | NP_001271323.1 | 12 | 1452 | |||
NM_001284398.1 | NP_001271327.1 | 11 | 1139 | |||
NM_011129.2 | NP_035259.1 | 12 | 1505 | |||
GenBank mRNA | AB078010.1 | - | 11 | 1103 | ||
AK133933.1 | - | 11 | 1107 | |||
AK135143.1 | - | 12 | 1506 | |||
BC055101.1 | - | 12 | 1452 | 6591016 | ||
X61452.1 | - | 12 | 1435 |