siRNA Details
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113559.2 | NP_001107031.2 | 10, 11 | 1552 | ||
NM_001243163.1 | NP_001230092.1 | 10, 11 | 1444 | |||
NM_011444.3 | NP_035574.2 | 11, 12 | 1696 | |||
GenBank mRNA | AB006330.1 | - | 11, 12 | 1608 | ||
AJ010604.1 | - | 10, 11 | 1434 | |||
AK015212.1 | - | 4, 5 | 505 | |||
AK029047.1 | - | 13, 14 | 1694 | |||
AK160154.1 | - | 4, 5 | 510 | |||
BC110478.2 | - | 10, 11 | 1436 | 40047865 | ||
KF793916.1 | - | 11, 12 | 1397 | |||
X65657.1 | - | 4, 5 | 481 |

More Information
Other Species Targeted

Panther Classification:
