Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNNT3 SIRNA Assays ›
          Gene Symbol:
          Tnnt3
          Gene Name:
          troponin T3, skeletal, fast
          Gene Aliases:
          fTnT
          Chromosome Location:
          Chr.7: 142498777 - 142516213 on Build GRCm38
          UniGene ID:
          Mm.389992
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 69643

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tnnt3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163664.1 NP_001157136.1 9, 10 240
          NM_001163665.1 NP_001157137.1 9, 10 240
          NM_001163666.1 NP_001157138.1 7, 8 213
          NM_001163667.1 NP_001157139.1 8, 9 228
          NM_001163668.1 NP_001157140.1 7, 8 189
          NM_001163669.1 NP_001157141.1 8, 9 201
          NM_001163670.1 NP_001157142.1 5, 6 156
          NM_011620.3 NP_035750.2 6, 7 168
          GenBank mRNA AA028290.1 - 8, 9 181
          AA754928.1 - 6, 7 145
          AA771596.1 - 7, 8 167
          AI173874.1 - 7, 8 205
          AI574395.1 - 9, 10 234
          AI746973.1 - 7, 8 208
          BC003747.1 - 9, 10 236 3594256
          BY766323.1 - 8, 9 218
          DV049834.1 - 6, 7 215
          L48988.1 - 3, 4 59
          L48989.1 - 3, 4 59
          L48990.1 - 6, 7 104
          L48991.1 - 3, 4 86
          L48992.1 - 5, 6 92
          L48993.1 - 5, 6 92
          L49018.1 - 3, 4 86
          L49466.1 - 8, 9 228
          L49467.1 - 9, 10 240
          L49468.1 - 7, 8 213
          L49469.1 - 7, 8 213
          L49470.1 - 8, 9 201
          L49471.1 - 7, 8 210
          L49472.1 - 7, 8 189
          U77779.1 - 6, 7 161

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001270677, NM_001270665, NM_001270675, NM_001270664, NM_001270676, and others.

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          regulation of muscle contraction
          regulation of striated muscle contraction
          metabolic process
          regulation of ATPase activity
          actin binding
          tropomyosin binding
          troponin C binding
          calcium-dependent ATPase activity
          troponin I binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0