siRNA Details
Gene Symbol: Vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011694.4 | NP_035824.1 | 7 | 640 | ||
GenBank mRNA | AK002740.1 | - | 7 | 700 | ||
AK146239.1 | - | 7 | 673 | |||
AK153094.1 | - | 7 | 642 | |||
AK167860.1 | - | 7 | 673 | |||
AK168225.1 | - | 7 | 649 | |||
AK168672.1 | - | 7 | 678 | |||
AK169160.1 | - | 7 | 678 | |||
AK169282.1 | - | 7 | 682 | |||
AK169354.1 | - | 7 | 678 | |||
AK169671.1 | - | 7 | 639 | |||
AK189657.1 | - | 2 | 181 | |||
BC092257.1 | - | 7 | 623 | 5124735 | ||
U30840.1 | - | 7 | 640 |

More Information
Other Species Targeted

Panther Classification:
