Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290648.1 | NP_001277577.1 | 3 | 308 | ||
NM_011831.2 | NP_035961.1 | 2 | 314 | |||
GenBank mRNA | AF054842.1 | - | 3 | 308 | ||
AF054843.1 | - | 2 | 278 | |||
AF076971.1 | - | 2 | 310 | |||
AF133817.1 | - | 2 | 317 | |||
BC010968.1 | - | 2 | 309 | 4217383 |