Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037940.4 | NP_001033029.1 | 4 | 467 | ||
NM_001037941.3 | NP_001033030.2 | 4 | 467 | |||
NM_001127367.1 | NP_001120839.1 | 4 | 467 | |||
NM_011847.4 | NP_035977.2 | 4 | 467 | |||
GenBank mRNA | AB028854.1 | - | 4 | 369 | ||
AF035962.1 | - | 4 | 367 | |||
AK005680.1 | - | 4 | 372 | |||
AK032039.1 | - | 4 | 376 | |||
AK133805.1 | - | 4 | 384 | |||
AK134387.1 | - | 4 | 426 | |||
AK144182.1 | - | 4 | 394 | |||
AK151219.1 | - | 4 | 374 | |||
AK151752.1 | - | 4 | 341 | |||
AK152626.1 | - | 4 | 381 | |||
AK152632.1 | - | 4 | 341 | |||
AK153091.1 | - | 4 | 347 | |||
AK169332.1 | - | 4 | 373 | |||
AK169767.1 | - | 4 | 338 | |||
AK180819.1 | - | 1 | 39 | |||
AK208008.1 | - | 4 | 300 | |||
AK216936.1 | - | 1 | 39 | |||
BC003702.1 | - | 4 | 349 | 3601332 | ||
BC083349.1 | - | 4 | 359 | 30541723 | ||
BG800394.1 | - | 4 | 357 | |||
DV660970.1 | - | 3 | 262 |