Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011875.4 | NP_036005.1 | 2 | 206 | ||
GenBank mRNA | AF107838.1 | - | 2 | 167 | ||
AK051078.1 | - | 2 | 197 | |||
AK075936.1 | - | 2 | 207 | |||
AK081160.1 | - | 2 | 204 | |||
AK145654.1 | - | 2 | 169 | |||
AK163017.1 | - | 2 | 201 | |||
AK167169.1 | - | 2 | 191 | |||
AK171504.1 | - | 2 | 187 | |||
AK206994.1 | - | 2 | 82 | |||
BC029989.1 | - | 2 | 146 | |||
BC139184.1 | - | 2 | 184 | 8862206 | ||
BC139187.1 | - | 2 | 184 | 8862209 |