siRNA Details
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127367.1 | NP_001120839.1 | 8 | 1031 | ||
NM_011847.4 | NP_035977.2 | 8 | 1034 | |||
GenBank mRNA | AB028854.1 | - | 8 | 936 | ||
AF035962.1 | - | 8 | 934 | |||
AK133805.1 | - | 8 | 951 | |||
AK134387.1 | - | 8 | 993 | |||
AK144182.1 | - | 8 | 955 | |||
AK151219.1 | - | 9 | 941 | |||
AK151752.1 | - | 8 | 908 | |||
AK152626.1 | - | 8 | 948 | |||
AK152632.1 | - | 8 | 908 | |||
AK169332.1 | - | 8 | 940 | |||
AK169767.1 | - | 8 | 902 | |||
BC003702.1 | - | 8 | 916 | 3601332 | ||
BC083349.1 | - | 8 | 926 | 30541723 |

More Information
Other Species Targeted

