Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112705.2 | NP_001106176.1 | 4 | 729 | ||
NM_001294329.1 | NP_001281258.1 | 4 | 729 | |||
NM_001294330.1 | NP_001281259.1 | 4 | 729 | |||
NM_001294331.1 | NP_001281260.1 | 5 | 838 | |||
NM_001294334.1 | NP_001281263.1 | 4 | 729 | |||
GenBank mRNA | AF045252.1 | - | 5 | 844 | ||
AF045253.1 | - | 3 | 176 | |||
AK014829.1 | - | 4 | 367 | |||
AK048492.1 | - | 1 | 1229 | |||
AK089925.1 | - | 4 | 257 | |||
AK165667.1 | - | 4 | 732 | |||
BC066198.1 | - | 4 | 274 |